Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198787.2 | NP_942082.2 | 8, 9 | 961 | ||
XM_006242096.2 | XP_006242158.1 | 8, 9 | 987 | |||
XM_006242098.2 | XP_006242160.1 | 8, 9 | 992 | |||
XM_006242099.2 | XP_006242161.1 | 8, 9 | 992 | |||
XM_008765761.1 | XP_008763983.1 | 8, 9 | 992 | |||
XM_008765762.1 | XP_008763984.1 | 8, 9 | 2693 | |||
XM_008765763.1 | XP_008763985.1 | 8, 9 | 987 | |||
GenBank mRNA | AB091009.2 | - | 6, 7 | 809 | ||
BC062060.1 | - | 8, 9 | 928 | 5599107 | ||
FQ212274.1 | - | 8, 9 | 959 |