Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317769.1 | NP_001304698.1 | 3, 4 | 412 | ||
NM_012108.3 | NP_036240.1 | 3, 4 | 412 | |||
XM_017008018.1 | XP_016863507.1 | 3, 4 | 412 | |||
GenBank mRNA | AB023483.1 | - | 3, 4 | 382 | ||
AK313676.1 | - | 3, 4 | 361 | |||
BC014958.1 | - | 3, 4 | 331 | 4851485 |