Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199104.2 | NP_954535.2 | 12 | 1855 | ||
XM_006231392.2 | XP_006231454.1 | 13 | 1892 | |||
XM_008760422.1 | XP_008758644.1 | 12 | 2400 | |||
XM_008760423.1 | XP_008758645.1 | 11 | 2379 | |||
GenBank mRNA | BC061741.1 | - | 12 | 1821 | 5599023 |