Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199117.2 | NP_954548.1 | 6 | 1951 | ||
XM_006242020.2 | XP_006242082.1 | 6 | 2360 | |||
XM_006242021.2 | XP_006242083.1 | 7 | 2133 | |||
XM_006242022.2 | XP_006242084.1 | 6 | 2508 | |||
XM_006242024.2 | XP_006242086.1 | 6 | 2166 | |||
XM_006242025.2 | XP_006242087.1 | 7 | 1829 | |||
XM_006242026.2 | XP_006242088.1 | 6 | 2062 | |||
GenBank mRNA | BC062392.1 | - | 6 | 1929 | 5621139 |