Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LANCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136574.1 | NP_001130046.1 | 10 | 4146 | ||
NM_001136575.1 | NP_001130047.1 | 10 | 4141 | |||
NM_006055.2 | NP_006046.1 | 10 | 4235 | |||
XM_005246243.2 | XP_005246300.1 | 10 | 4256 | |||
GenBank mRNA | Y11395.1 | - | 10 | 4174 |