Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012852.1 | NP_036984.1 | 1 | 824 | ||
XM_008764208.1 | XP_008762430.1 | 2 | 1617 |
Set Membership: |
![]() |