Gene Symbol: FBXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012168.5 | NP_036300.2 | 6 | 1535 | ||
GenBank mRNA | AF187318.1 | - | 6 | 1383 | ||
BC025233.1 | - | 6 | 1119 | 5090925 | ||
BC096747.1 | - | 6 | 1135 | 5259298 |