Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012862.1 | NP_036994.1 | 4 | 341 | ||
GenBank mRNA | BC086394.1 | - | 4 | 373 | 7303359 | |
FQ221152.1 | - | 2 | 179 | |||
FQ221407.1 | - | 2 | 179 | |||
FQ224403.1 | - | 4 | 398 | |||
FQ229124.1 | - | 2 | 179 | |||
J03026.1 | - | 4 | 341 |