Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012869.1 | NP_037001.1 | 2 | 363 | ||
XM_006253013.2 | XP_006253075.1 | 2 | 218 | |||
XM_006253014.2 | XP_006253076.1 | 3 | 408 | |||
GenBank mRNA | AF044264.1 | - | 3 | 421 | ||
U56078.1 | - | 1 | 180 | |||
U66274.1 | - | 2 | 363 |
Set Membership: |
![]() |