Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012880.1 | NP_037012.1 | 2 | 249 | ||
XM_006251030.2 | XP_006251092.1 | 2 | 295 | |||
GenBank mRNA | BC061861.1 | - | 2 | 200 | 5598594 | |
X68041.1 | - | 2 | 224 | |||
X94371.1 | - | 2 | 217 | |||
Z24721.1 | - | 2 | 249 |