Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sstr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012882.2 | NP_037014.2 | 2 | 1237 | ||
GenBank mRNA | L04535.1 | - | 2 | 1240 |
Set Membership: |
![]() |