Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012939.1 | NP_037071.1 | 3, 4 | 221 | ||
XM_008766440.1 | XP_008764662.1 | 3, 4 | 272 | |||
GenBank mRNA | BC085352.1 | - | 3, 4 | 236 | 7115929 | |
FQ210730.1 | - | 3, 4 | 231 | |||
FQ213183.1 | - | 3, 4 | 231 | |||
FQ219177.1 | - | 3, 4 | 231 | |||
FQ219285.1 | - | 3, 4 | 231 | |||
FQ219548.1 | - | 3, 4 | 231 | |||
FQ219611.1 | - | 3, 4 | 249 | |||
FQ223707.1 | - | 3, 4 | 227 | |||
FQ228686.1 | - | 3, 4 | 291 | |||
FQ228906.1 | - | 3, 4 | 238 | |||
FQ228938.1 | - | 3, 4 | 228 | |||
FQ228956.1 | - | 3, 4 | 258 | |||
M38135.1 | - | 3, 4 | 215 | |||
Y00708.1 | - | 3, 4 | 221 |