Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013011.3 | NP_037143.2 | 2 | 280 | ||
XM_006241529.2 | XP_006241591.1 | 3 | 833 | |||
GenBank mRNA | BC070941.1 | - | 2 | 280 | 7099670 | |
BC094305.1 | - | 2 | 585 | 7313193 | ||
D17615.1 | - | 2 | 347 | |||
D30740.1 | - | 2 | 260 | |||
L07913.1 | - | 1 | 49 | |||
U37252.1 | - | 2 | 281 |