Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012256.3 | NP_036388.2 | 3 | 558 | ||
XM_005250055.2 | XP_005250112.2 | 3 | 556 | |||
XM_011516581.2 | XP_011514883.2 | 3 | 546 | |||
XM_011516582.1 | XP_011514884.1 | 3 | 478 | |||
XM_017012646.1 | XP_016868135.1 | 3 | 559 | |||
XM_017012647.1 | XP_016868136.1 | 4 | 512 | |||
XM_017012648.1 | XP_016868137.1 | 3 | 801 | |||
GenBank mRNA | AK074821.1 | - | 4 | 561 | ||
AK315086.1 | - | 3 | 567 | |||
BC022785.2 | - | 3 | 525 | 3849803 |