Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013065.2 | NP_037197.1 | 8 | 2372 | ||
XM_008764505.1 | XP_008762727.1 | 9 | 2310 | |||
GenBank mRNA | BC062033.1 | - | 8 | 2372 | 5623630 | |
D90164.1 | - | 8 | 2152 | |||
FQ212294.1 | - | 9 | 2310 | |||
FQ226372.1 | - | 1 | 511 | |||
FQ227415.1 | - | 1 | 448 | |||
S78218.1 | - | 8 | 2139 |