Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Map2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013066.1 | NP_037198.1 | 5 | 3014 | ||
XM_006245100.2 | XP_006245162.1 | 5 | 7163 | |||
XM_008767197.1 | XP_008765419.1 | 5 | 3194 | |||
XM_008767198.1 | XP_008765420.1 | 5 | 3194 | |||
XM_008767199.1 | XP_008765421.1 | 5 | 3191 | |||
XM_008767200.1 | XP_008765422.1 | 5 | 3194 | |||
XM_008767201.1 | XP_008765423.1 | 5 | 3191 | |||
XM_008767202.1 | XP_008765424.1 | 5 | 3182 | |||
XM_008767203.1 | XP_008765425.1 | 5 | 3194 | |||
XM_008767204.1 | XP_008765426.1 | 4 | 2948 | |||
XM_008767205.1 | XP_008765427.1 | 4 | 2936 | |||
XM_008767206.1 | XP_008765428.1 | 5 | 2982 | |||
GenBank mRNA | X51842.1 | - | 5 | 2983 | ||
X54100.1 | - | 5 | 3014 |