Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013081.2 | NP_037213.1 | 4 | 845 | ||
XM_008765546.1 | XP_008763768.1 | 6 | 974 | |||
GenBank mRNA | AF020777.1 | - | 4 | 844 |