Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177593.1 | NP_001171064.1 | 9 | 1131 | ||
NM_013088.2 | NP_037220.2 | 9 | 1131 | |||
GenBank mRNA | D83016.1 | - | 3 | 180 | ||
U05963.1 | - | 9 | 1062 | |||
U09307.1 | - | 9 | 1056 | |||
U57499.1 | - | 1 | 35 |