Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751127.3 | XP_003751175.1 | 13 | 1902 |
Gene Symbol: Cyp3a23/3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D13912.1 | - | 13 | 1872 | ||
D29967.1 | - | 1874 | ||||
FQ211106.1 | - | 1887 | ||||
FQ218135.1 | - | 2607 | ||||
L24207.1 | - | 1863 | ||||
X96721.1 | - | 1874 |