Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013108.2 | NP_037240.2 | 3 | 1759 | ||
XM_008771315.1 | XP_008769537.1 | 2 | 2288 | |||
GenBank mRNA | M74716.1 | - | 3 | 1758 | ||
S56481.1 | - | 3 | 1550 |
Set Membership: |
![]() |