Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013123.3 | NP_037255.3 | 3 | 184 | ||
XM_006244746.2 | XP_006244808.1 | 4 | 2293 | |||
XM_006244747.2 | XP_006244809.1 | 4 | 550 | |||
XM_006244748.2 | XP_006244810.1 | 3 | 454 | |||
XM_006244749.2 | XP_006244811.1 | 5 | 373 | |||
XM_006244750.2 | XP_006244812.1 | 5 | 2365 | |||
XM_006244751.2 | XP_006244813.1 | 4 | 284 | |||
XM_006244752.2 | XP_006244814.1 | 4 | 526 | |||
XM_006244753.2 | XP_006244815.1 | 4 | 1645 | |||
XM_006244754.2 | XP_006244816.1 | 3 | 2197 | |||
XM_008766993.1 | XP_008765215.1 | 4 | 301 | |||
GenBank mRNA | AB112436.1 | - | 5 | 318 | ||
AB112437.1 | - | 3 | 184 | |||
AB112438.1 | - | 4 | 256 | |||
M95578.1 | - | 4 | 505 | |||
U14010.1 | - | 2 | 133 |