Gene Symbol: Cald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013146.2 | NP_037278.1 | 3 | 413 | ||
XM_006236260.2 | XP_006236322.1 | 3 | 1963 | |||
XM_006236261.2 | XP_006236323.1 | 4 | 484 | |||
XM_006236262.2 | XP_006236324.1 | 4 | 637 | |||
XM_008762763.1 | XP_008760985.1 | 4 | 487 |