Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DBH SIRNA Assays ›
Gene Symbol:
Dbh
Gene Name:
dopamine beta-hydroxylase (dopamine beta-monooxygenase)
Gene Aliases:
DOPBHY
Chromosome Location:
Chr.3: 5709236 - 5731895 on Build Rnor_6.0
UniGene ID:
Rn.87166
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
197895
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dbh
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_013158.2
NP_037290.2
1
185
Back To Top
More Information
Panther Classification:
Molecular Function -
hydroxylase
oxidoreductase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
blood vessel remodeling
response to amphetamine
leukocyte mediated immunity
catecholamine metabolic process
memory
locomotory behavior
associative learning
visual learning
response to iron ion
response to ozone
multicellular organism aging
response to organic cyclic compound
response to estradiol
social behavior
response to isolation stress
response to immobilization stress
regulation of cell proliferation
homoiothermy
dopamine catabolic process
norepinephrine biosynthetic process
response to drug
glucose homeostasis
fear response
maternal behavior
response to peptide hormone
response to ethanol
positive regulation of vasoconstriction
octopamine metabolic process
response to copper ion
behavioral response to ethanol
response to pain
leukocyte migration
sensory perception of taste
oxidation-reduction process
bone development
cellular response to manganese ion
cellular response to nicotine
regulation of extrinsic apoptotic signaling pathway
dopamine beta-monooxygenase activity
copper ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced ascorbate as one donor, and incorporation of one atom of oxygen
L-ascorbic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-b9vj9:80/10.239.164.80:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0