Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001556.2 | NP_001001556.1 | 6, 7 | 811 | ||
NM_001289030.1 | NP_001275959.1 | 7, 8 | 1043 | |||
NM_001289031.1 | NP_001275960.1 | 6, 7 | 896 | |||
NM_002044.3 | NP_002035.1 | 6, 7 | 913 | |||
XM_005254279.3 | XP_005254336.1 | 6, 7 | 702 | |||
XM_005254280.3 | XP_005254337.1 | 6, 7 | 793 | |||
XM_006720461.3 | XP_006720524.1 | 6, 7 | 702 | |||
XM_006720462.3 | XP_006720525.1 | 6, 7 | 683 | |||
XM_011521441.2 | XP_011519743.1 | 7, 8 | 906 | |||
XM_017022064.1 | XP_016877553.1 | 4, 5 | 759 | |||
XM_017022065.1 | XP_016877554.1 | 7, 8 | 958 | |||
XM_017022066.1 | XP_016877555.1 | 7, 8 | 979 | |||
GenBank mRNA | AF461816.1 | - | 6, 7 | 791 | ||
AK098049.1 | - | 7, 8 | 767 | |||
AK301361.1 | - | 6, 7 | 666 | |||
AK316440.1 | - | 7, 8 | 777 | |||
AL832745.1 | - | 6, 7 | 790 | |||
BC005141.2 | - | 6, 7 | 681 | 3504930 | ||
BQ921903.1 | - | 6, 7 | 624 | |||
BT006901.1 | - | 6, 7 | 606 | |||
EB386877.1 | - | 4, 5 | 442 | |||
M84443.1 | - | 6, 7 | 626 |