Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017103.2 | NP_058799.2 | 4 | 1031 | ||
GenBank mRNA | BC081815.1 | - | 4 | 1031 | 7110428 | |
L46816.1 | - | 1046 |