Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017161.1 | NP_058857.1 | 2 | 916 | ||
GenBank mRNA | BC161854.1 | - | 2 | 924 | 9024632 | |
M91466.1 | - | 2 | 916 |
Set Membership: |
![]() |