Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763523.1 | XP_008761745.1 | 3 | 2070 | ||
GenBank mRNA | U00442.1 | - | 4 | 2127 |
Set Membership: |
G-protein coupled receptor |