Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160154.1 | NP_001153626.1 | 8 | 846 | ||
NM_012214.2 | NP_036346.1 | 11 | 1430 | |||
GenBank mRNA | AB000616.1 | - | 10 | 1251 | ||
AK291533.1 | - | 10 | 1240 | |||
AK304393.1 | - | 8 | 846 | |||
AK314024.1 | - | 11 | 1390 | |||
BC127107.1 | - | 10 | 1328 | 40124731 | ||
BC127108.1 | - | 10 | 1328 | 40124741 |