Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aadat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017193.1 | NP_058889.1 | 2, 3 | 350 | ||
XM_006253067.2 | XP_006253129.1 | 2, 3 | 438 | |||
GenBank mRNA | BC078864.1 | - | 2, 3 | 346 | 7109210 | |
Z50144.1 | - | 2, 3 | 350 |