Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177321.1 | NP_001170792.1 | 3 | 267 | ||
NM_017200.1 | NP_058896.1 | 2 | 195 | |||
XM_006234440.2 | XP_006234502.1 | 3 | 490 | |||
XM_006234441.2 | XP_006234503.1 | 3 | 1511 | |||
XM_006234444.2 | XP_006234506.1 | 3 | 1524 | |||
XM_008761980.1 | XP_008760202.1 | 3 | 497 | |||
XM_008761981.1 | XP_008760203.1 | 3 | 261 | |||
XM_008761982.1 | XP_008760204.1 | 3 | 491 | |||
XM_008761983.1 | XP_008760205.1 | 3 | 1514 | |||
XM_008761984.1 | XP_008760206.1 | 3 | 1527 | |||
XM_008761985.1 | XP_008760207.1 | 3 | 503 | |||
XM_008761986.1 | XP_008760208.1 | 3 | 497 | |||
GenBank mRNA | BC087020.1 | - | 3 | 267 | 7192382 | |
D10926.1 | - | 2 | 195 |