Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NT5C2 SIRNA Assays ›
Gene Symbol:
NT5C2
Gene Name:
5'-nucleotidase, cytosolic II
Gene Aliases:
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location:
Chr.10: 103088017 - 103193306 on Build GRCh38
UniGene ID:
Hs.97439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19830
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NT5C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134373.2
NP_001127845.1
10
823
NM_012229.4
NP_036361.1
11
921
XM_005269632.4
XP_005269689.1
11
832
XM_005269633.4
XP_005269690.1
12
925
XM_005269634.4
XP_005269691.1
12
957
XM_005269635.4
XP_005269692.1
11
1147
XM_005269636.4
XP_005269693.1
11
933
XM_005269637.4
XP_005269694.1
10
1651
XM_005269638.4
XP_005269695.1
10
2952
XM_005269639.4
XP_005269696.1
9
676
XM_005269640.4
XP_005269697.1
12
948
XM_005269641.4
XP_005269698.1
11
850
XM_005269642.4
XP_005269699.1
10
775
XM_005269643.4
XP_005269700.1
9
715
XM_005269644.4
XP_005269701.1
13
972
XM_005269645.4
XP_005269702.1
12
874
XM_005269646.4
XP_005269703.1
11
800
XM_006717721.3
XP_006717784.1
11
874
XM_006717723.3
XP_006717786.1
9
671
XM_011539537.2
XP_011537839.1
12
957
XM_017015947.1
XP_016871436.1
9
701
XM_017015948.1
XP_016871437.1
12
1057
XM_017015949.1
XP_016871438.1
12
1042
XM_017015950.1
XP_016871439.1
10
758
XM_017015951.1
XP_016871440.1
9
684
XM_017015952.1
XP_016871441.1
11
981
XM_017015953.1
XP_016871442.1
12
1376
XM_017015954.1
XP_016871443.1
11
1315
XM_017015955.1
XP_016871444.1
11
1002
XM_017015956.1
XP_016871445.1
11
1208
XM_017015957.1
XP_016871446.1
13
1066
XM_017015958.1
XP_016871447.1
14
1185
XM_017015959.1
XP_016871448.1
10
776
XM_017015960.1
XP_016871449.1
11
739
XM_017015961.1
XP_016871450.1
11
1134
XM_017015962.1
XP_016871451.1
11
968
XM_017015963.1
XP_016871452.1
11
1072
XM_017015964.1
XP_016871453.1
11
1051
XM_017015965.1
XP_016871454.1
10
928
XM_017015966.1
XP_016871455.1
10
928
XM_017015967.1
XP_016871456.1
10
863
XM_017015968.1
XP_016871457.1
11
856
XM_017015969.1
XP_016871458.1
11
950
XM_017015970.1
XP_016871459.1
9
1593
XM_017015971.1
XP_016871460.1
8
637
XM_017015972.1
XP_016871461.1
12
891
XM_017015973.1
XP_016871462.1
13
915
XM_017015974.1
XP_016871463.1
12
915
XM_017015975.1
XP_016871464.1
10
756
XM_017015976.1
XP_016871465.1
10
762
GenBank mRNA
AK127670.1
-
11
912
AK291667.1
-
10
789
AK295593.1
-
9
965
AK312517.1
-
10
789
BC001595.1
-
10
872
3543103
BX648078.1
-
12
2723
D38524.1
-
10
811
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide catabolic process
phosphorylation
dephosphorylation
drug metabolic process
IMP metabolic process
adenosine metabolic process
nucleotide binding
protein binding
5'-nucleotidase activity
metal ion binding
nucleoside phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0