Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pctp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017225.2 | NP_058921.1 | 6 | 601 | ||
XM_006247124.2 | XP_006247186.1 | 6 | 587 | |||
XM_006247125.2 | XP_006247187.1 | 5 | 601 | |||
XM_008767982.1 | XP_008766204.1 | 8 | 808 | |||
XM_008767983.1 | XP_008766205.1 | 8 | 762 | |||
GenBank mRNA | AF040261.1 | - | 6 | 545 | ||
BC078854.1 | - | 6 | 606 | 7113616 |