Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017295.1 | NP_058991.1 | 4 | 425 | ||
GenBank mRNA | X51992.1 | - | 4 | 452 |