Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019141.2 | NP_062014.2 | 5, 6 | 497 | ||
GenBank mRNA | CB717588.1 | - | 4, 5 | 222 | ||
L14463.1 | - | 5, 6 | 493 |