Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTA1 SIRNA Assays ›
Gene Symbol:
Acta1
Gene Name:
actin, alpha 1, skeletal muscle
Gene Aliases:
-
Chromosome Location:
Chr.19: 56674072 - 56677084 on Build Rnor_6.0
UniGene ID:
Rn.82732
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
198747
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acta1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019212.2
NP_062085.1
3
454
GenBank mRNA
BC061974.1
-
3
454
5623909
FQ214583.1
-
3
463
FQ214598.1
-
3
463
FQ214600.1
-
3
463
FQ214603.1
-
3
463
FQ214623.1
-
3
463
FQ214640.1
-
3
463
FQ214659.1
-
3
465
FQ214676.1
-
3
463
FQ214680.1
-
3
463
FQ214697.1
-
3
463
FQ214734.1
-
3
463
FQ214743.1
-
3
463
FQ214756.1
-
3
463
FQ214761.1
-
3
462
FQ214779.1
-
3
463
FQ214785.1
-
3
461
FQ214786.1
-
3
463
FQ214807.1
-
3
463
FQ214814.1
-
3
463
FQ214822.1
-
3
463
FQ214831.1
-
3
463
FQ214832.1
-
3
463
FQ214839.1
-
3
466
FQ214841.1
-
3
463
FQ214842.1
-
3
463
FQ214857.1
-
3
467
FQ214858.1
-
3
463
FQ214859.1
-
3
463
FQ214860.1
-
3
463
FQ214862.1
-
3
463
FQ214866.1
-
3
463
FQ214883.1
-
3
462
FQ214904.1
-
3
463
FQ214905.1
-
3
462
FQ214916.1
-
3
463
FQ214936.1
-
3
463
FQ214948.1
-
3
463
FQ214949.1
-
3
463
FQ214957.1
-
3
463
FQ214969.1
-
3
463
FQ214975.1
-
3
465
FQ214976.1
-
3
463
FQ214978.1
-
3
463
FQ214987.1
-
3
463
FQ214990.1
-
3
463
FQ214993.1
-
3
463
FQ215012.1
-
3
463
FQ215025.1
-
3
463
FQ215038.1
-
3
463
FQ215044.1
-
3
463
FQ215058.1
-
3
463
FQ215089.1
-
3
463
FQ215112.1
-
3
463
FQ215128.1
-
3
465
FQ215149.1
-
3
463
FQ215153.1
-
3
463
FQ215158.1
-
3
461
FQ215173.1
-
3
463
FQ215204.1
-
3
461
FQ215212.1
-
3
463
FQ215215.1
-
3
463
FQ215225.1
-
3
463
FQ215241.1
-
3
463
FQ215249.1
-
3
464
FQ215253.1
-
3
463
FQ215256.1
-
3
463
FQ215275.1
-
3
463
FQ215277.1
-
3
464
FQ215280.1
-
3
463
FQ215282.1
-
3
463
FQ215298.1
-
3
465
FQ215324.1
-
3
463
FQ215333.1
-
3
463
FQ215342.1
-
3
463
FQ215346.1
-
3
463
FQ215366.1
-
3
463
FQ215380.1
-
3
463
FQ215381.1
-
3
463
FQ215393.1
-
3
463
FQ215396.1
-
3
463
FQ215398.1
-
3
463
FQ215402.1
-
3
463
FQ215403.1
-
3
463
FQ215405.1
-
3
463
FQ215414.1
-
3
465
FQ215431.1
-
3
463
FQ215433.1
-
3
462
FQ215434.1
-
3
465
FQ215438.1
-
3
463
FQ215446.1
-
3
463
FQ215449.1
-
3
465
FQ215454.1
-
3
463
FQ215464.1
-
3
463
FQ215465.1
-
3
463
FQ215468.1
-
3
463
FQ215470.1
-
3
463
FQ215474.1
-
3
463
FQ215483.1
-
3
463
FQ215488.1
-
3
463
FQ215518.1
-
3
472
FQ215523.1
-
3
463
FQ215537.1
-
3
465
FQ215554.1
-
3
463
FQ215567.1
-
3
463
FQ215572.1
-
3
463
FQ215593.1
-
3
463
FQ215618.1
-
3
463
FQ215628.1
-
3
463
FQ215631.1
-
1
77
FQ215655.1
-
3
461
FQ215659.1
-
3
462
FQ215673.1
-
3
463
FQ215674.1
-
3
463
FQ215690.1
-
3
463
FQ215692.1
-
3
463
FQ215785.1
-
3
463
FQ215790.1
-
3
465
FQ215799.1
-
3
463
FQ215809.1
-
3
463
FQ215813.1
-
3
463
FQ215835.1
-
3
463
FQ215836.1
-
3
463
FQ215839.1
-
3
463
FQ215840.1
-
3
463
FQ215841.1
-
3
463
FQ215851.1
-
3
463
FQ215852.1
-
3
461
FQ215864.1
-
3
464
FQ215866.1
-
3
465
FQ215888.1
-
3
462
FQ215893.1
-
3
464
FQ215896.1
-
3
463
FQ215907.1
-
3
465
FQ215924.1
-
1
155
FQ215950.1
-
3
463
FQ215964.1
-
3
463
FQ215972.1
-
3
463
FQ216016.1
-
3
463
FQ216035.1
-
3
463
FQ216043.1
-
3
463
FQ216050.1
-
3
463
FQ216066.1
-
3
461
FQ216138.1
-
3
463
FQ216139.1
-
3
463
FQ216168.1
-
3
461
FQ216196.1
-
3
463
FQ216224.1
-
3
463
FQ216246.1
-
3
463
FQ216276.1
-
3
463
FQ216278.1
-
3
465
FQ216287.1
-
3
461
FQ216291.1
-
3
463
FQ216293.1
-
3
463
FQ216305.1
-
3
463
FQ216329.1
-
3
463
FQ216333.1
-
3
463
FQ216350.1
-
3
463
FQ216352.1
-
3
463
FQ216360.1
-
3
463
FQ216369.1
-
3
465
FQ216403.1
-
3
463
FQ216430.1
-
3
463
FQ216509.1
-
3
461
FQ216586.1
-
3
463
FQ216603.1
-
3
463
FQ216606.1
-
3
463
FQ216607.1
-
3
459
FQ216610.1
-
3
461
FQ216620.1
-
3
463
FQ216635.1
-
3
461
FQ216639.1
-
3
463
FQ216644.1
-
3
463
FQ216664.1
-
3
461
FQ216675.1
-
3
463
FQ216676.1
-
3
463
FQ216692.1
-
3
463
FQ216703.1
-
3
463
FQ216713.1
-
3
463
FQ216721.1
-
3
463
FQ216723.1
-
3
463
FQ216725.1
-
3
459
FQ216739.1
-
3
463
FQ216762.1
-
3
461
FQ216767.1
-
3
463
FQ216776.1
-
3
463
FQ216777.1
-
3
463
FQ216793.1
-
3
463
FQ216799.1
-
3
461
FQ216801.1
-
3
463
FQ216814.1
-
3
457
FQ216819.1
-
3
461
FQ216916.1
-
3
463
FQ216918.1
-
1
106
FQ216961.1
-
3
463
FQ216994.1
-
3
463
FQ217061.1
-
3
465
FQ217073.1
-
3
466
FQ217091.1
-
3
462
FQ217114.1
-
3
463
FQ217122.1
-
3
463
FQ217167.1
-
3
463
FQ217173.1
-
3
463
FQ217188.1
-
3
463
FQ217194.1
-
3
463
FQ217280.1
-
3
583
FQ217327.1
-
3
463
FQ217329.1
-
3
463
FQ217347.1
-
3
463
FQ217353.1
-
3
463
FQ217367.1
-
3
463
FQ217397.1
-
3
463
FQ217441.1
-
3
463
FQ217443.1
-
3
465
FQ217475.1
-
3
463
FQ217525.1
-
3
463
FQ217535.1
-
3
463
FQ217572.1
-
3
463
FQ217593.1
-
3
463
FQ217602.1
-
3
463
FQ217615.1
-
3
463
FQ217629.1
-
3
463
FQ217641.1
-
3
460
FQ217762.1
-
3
463
FQ217826.1
-
3
463
FQ217884.1
-
3
461
FQ217893.1
-
3
463
FQ217905.1
-
3
463
FQ217908.1
-
3
463
FQ217919.1
-
3
463
FQ217937.1
-
3
463
FQ217969.1
-
3
465
FQ217990.1
-
3
463
FQ218019.1
-
3
463
FQ218034.1
-
3
463
FQ223563.1
-
3
463
FQ223681.1
-
3
465
FQ223686.1
-
3
463
FQ223702.1
-
3
463
FQ223727.1
-
1
161
FQ223898.1
-
3
463
FQ223914.1
-
3
463
FQ223926.1
-
3
463
FQ223929.1
-
3
463
FQ224009.1
-
3
463
FQ224082.1
-
3
463
FQ224141.1
-
3
464
FQ224167.1
-
3
463
FQ224359.1
-
3
463
FQ224377.1
-
3
463
FQ224419.1
-
3
463
FQ224691.1
-
3
461
FQ224840.1
-
3
461
FQ224842.1
-
3
463
FQ224862.1
-
3
463
FQ224869.1
-
3
463
FQ224886.1
-
3
463
FQ224890.1
-
3
461
FQ224902.1
-
3
463
FQ224929.1
-
3
461
FQ224936.1
-
3
463
FQ224942.1
-
3
465
FQ224948.1
-
3
465
FQ224962.1
-
3
463
FQ224971.1
-
3
463
Back To Top
More Information
Panther Classification:
Molecular Function -
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
response to mechanical stimulus
response to extracellular stimulus
response to lithium ion
cell growth
skeletal muscle thin filament assembly
skeletal muscle fiber adaptation
response to steroid hormone
skeletal muscle fiber development
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline