Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000685.4 | NP_000676.1 | 3 | 1056 | ||
NM_004835.4 | NP_004826.5 | 3 | 1030 | |||
NM_009585.3 | NP_033611.1 | 2 | 972 | |||
NM_031850.3 | NP_114038.4 | 4 | 1114 | |||
NM_032049.3 | NP_114438.2 | 3 | 930 | |||
GenBank mRNA | AK291541.1 | - | 3 | 1025 | ||
D13814.1 | - | 1 | 907 | |||
M87290.1 | - | 2 | 948 | |||
M93394.1 | - | 3 | 840 | |||
S77410.1 | - | 3 | 1002 | |||
X65699.1 | - | 2 | 905 |
Set Membership: |
![]() |