Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF020683.1 | - | 5 | 579 | ||
FM059586.1 | - | 5 | 491 | |||
FM081441.1 | - | 5 | 517 |