Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019269.1 | NP_062142.1 | 10 | 2143 | ||
GenBank mRNA | AB017260.1 | - | 10 | 2143 | ||
AF110416.1 | - | 10 | 2143 | |||
AJ001933.1 | - | 10 | 2129 | |||
BC166421.1 | - | 9 | 1892 | 9089728 |
Gene Symbol: LOC303140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767743.1 | XP_008765965.1 | 10 | 2791 | ||
GenBank mRNA | DQ119106.1 | - | 8 | 2008 |