Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hspa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019271.2 | NP_062144.2 | 4 | 607 | ||
GenBank mRNA | AF006617.1 | - | 4 | 607 | ||
BC070954.1 | - | 4 | 607 | 7103134 |