Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cacna1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020084.3 | NP_064469.3 | 2 | 302 | ||
GenBank mRNA | AF086827.2 | - | 567 | |||
AF290214.4 | - | 2 | 302 |