Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012400.3 | NP_036532.1 | 3 | 325 | ||
GenBank mRNA | AF112982.1 | - | 3 | 285 | ||
AF188625.1 | - | 3 | 293 | |||
AK290406.1 | - | 3 | 286 | |||
AK301396.1 | - | 3 | 325 | |||
AK311903.1 | - | 3 | 295 | |||
BC025706.1 | - | 3 | 274 | 5223912 | ||
DC295429.1 | - | 3 | 325 |