Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022676.3 | NP_073167.1 | 4 | 347 | ||
XM_006242437.2 | XP_006242499.1 | 4 | 359 | |||
GenBank mRNA | AJ276593.1 | - | 4 | 220 | ||
AY648296.1 | - | 4 | 347 | |||
BC078820.1 | - | 4 | 348 | 7127505 | ||
FQ217772.1 | - | 4 | 316 | |||
J05592.1 | - | 4 | 219 |