Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030997.1 | NP_112259.1 | 3 | 1232 | ||
XM_006249137.2 | XP_006249199.1 | 3 | 1253 | |||
XM_006249138.2 | XP_006249200.1 | 2 | 1102 | |||
XM_008769134.1 | XP_008767356.1 | 2 | 1305 | |||
GenBank mRNA | M60525.1 | - | 3 | 1152 | ||
M74223.1 | - | 2 | 1098 |
Gene Symbol: LOC100909521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257726.2 | XP_006257788.1 | 1305 |