Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031579.2 | NP_113767.1 | 6 | 1143 | ||
XM_006244670.2 | XP_006244732.1 | 6 | 1025 | |||
XM_006244671.2 | XP_006244733.1 | 6 | 1008 | |||
GenBank mRNA | AY062269.1 | - | 5 | 426 | ||
BC081772.1 | - | 5 | 773 | 7124031 | ||
BC097307.1 | - | 6 | 1143 | 7446277 | ||
L27843.1 | - | 6 | 972 |
Gene Symbol: LOC100362117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773245.1 | XP_008771467.1 | 3 | 367 |
Gene Symbol: LOC103693189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766779.1 | XP_008765001.1 | 5 | 942 |