Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199897.1 | NP_001186826.1 | 3 | 781 | ||
NM_012261.3 | NP_036393.1 | 4 | 913 | |||
GenBank mRNA | AK295336.1 | - | 3 | 781 | ||
AK316114.1 | - | 3 | 781 | |||
AL121740.1 | - | 4 | 620 | |||
BC050727.1 | - | 4 | 616 | 6044324 |