Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031621.1 | NP_113809.1 | 1 | 371 | ||
GenBank mRNA | JX215366.1 | - | 1192 | |||
U24652.1 | - | 3 | 218 | |||
U24653.1 | - | 3 | 218 | |||
U24654.1 | - | 3 | 218 | |||
U24655.1 | - | 1 | 371 |