Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031696.3 | NP_001026866.1 | 11, 12 | 1588 | ||
NM_001291311.1 | NP_001278240.1 | 11, 12 | 1522 | |||
NM_012268.3 | NP_036400.2 | 11, 12 | 1812 | |||
XM_005258704.1 | XP_005258761.1 | 11, 12 | 1733 | |||
XM_005258707.4 | XP_005258764.1 | 11, 12 | 1515 | |||
XM_005258708.3 | XP_005258765.1 | 10, 11 | 1408 | |||
XM_005258709.4 | XP_005258766.1 | 10, 11 | 1396 | |||
XM_005258710.4 | XP_005258767.1 | 10, 11 | 1437 | |||
XM_006723122.1 | XP_006723185.1 | 11, 12 | 1900 | |||
XM_011526692.1 | XP_011524994.1 | 11, 12 | 1895 | |||
XM_011526693.1 | XP_011524995.1 | 11, 12 | 1615 | |||
XM_017026546.1 | XP_016882035.1 | 12, 13 | 1831 | |||
XM_017026547.1 | XP_016882036.1 | 11, 12 | 1399 | |||
XM_017026548.1 | XP_016882037.1 | 11, 12 | 1550 | |||
XM_017026549.1 | XP_016882038.1 | 11, 12 | 1653 | |||
GenBank mRNA | AK293690.1 | - | 11, 12 | 1256 | ||
BC000553.1 | - | 7, 8 | 1058 | |||
BC036327.1 | - | 11, 12 | 1313 | 5189261 | ||
BC096820.1 | - | 11, 12 | 1511 | 30558682 | ||
U60644.1 | - | 11, 12 | 1518 |