Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053294.4 | NP_445746.3 | 3 | 2473 | ||
GenBank mRNA | L08102.1 | - | 2 | 2045 | ||
M91214.2 | - | 2 | 2329 | |||
S47609.1 | - | 2 | 2123 |
Set Membership: |
![]() |