Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053296.1 | NP_445748.1 | 1 | 113 | ||
XM_006232506.2 | XP_006232568.1 | 2 | 259 | |||
GenBank mRNA | AJ310839.1 | - | 1 | 113 | ||
FQ212388.1 | - | 2 | 264 |