Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ip6k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053316.2 | NP_445768.1 | 6 | 1558 | ||
XM_006243839.1 | XP_006243901.1 | 6 | 1760 | |||
GenBank mRNA | AB049151.1 | - | 6 | 1609 | ||
BC078702.1 | - | 6 | 1558 | 7132706 | ||
FQ211545.1 | - | 6 | 1706 |